Learn More
Cetirizine Dihydrochloride 1.0 mg/ml in Water (as free base), LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Cetirizine Dihydrochloride 1.0 mg/ml in Water (as free base)
Specifications
Specifications
| Chemical Name or Material | Cetirizine Dihydrochloride |
| CAS | 83881-52-1 |
| Analyte or Component Names | Cetirizine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H25 Cl N2 O3 . 2 Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Cetirizine Dihydrochloride, Acetic acid, [2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9 CI), Alercet, Alerid, Alerlisin, Cesta, Cetirizine hydrochloride, Cetrine, Cetrizet, Cistamine, Formistin, Histazine, Nosemin, P 071, Reactine, Riztec, Ryzen, Sancotec, Selitex, Triz, UCB-P 071, Virlix, Zeran, Zirtec, Zirtek, Zirtin, Zyrlex, Zyrtec, Zyrzine, Cetiriz, Cetirizine dihydrochloride, Cetrak, Epirizine, Histazine-1, Tomazine |
| InChI Formula | InChI=1 S/C21H25ClN2O3.2 ClH/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26;;/h1-9,21 H,10-16H2,(H,25,26);2*1 H |
| SMILES | Cl.Cl.OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC Name | 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride |
| Show More |