Learn More
Cetirizine.2HCl, 1mg/ml in Acetonitrile/Water : 1/1 (as free base), Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Cetirizine.2HCl, 1mg/ml in Acetonitrile/Water : 1/1 (as free base)
Specifications
Specifications
| Chemical Name or Material | Cetirizine Dihydrochloride |
| CAS | 83881-52-1 |
| Analyte or Component Names | Cetirizine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H25 Cl N2 O3 . 2 Cl H |
| Concentration | 1 mg/mL |
| Synonym | Cetirizine Dihydrochloride, Acetic acid, [2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9 CI), Alercet, Alerid, Alerlisin, Cesta, Cetirizine hydrochloride, Cetrine, Cetrizet, Cistamine, Formistin, Histazine, Nosemin, P 071, Reactine, Riztec, Ryzen, Sancotec, Selitex, Triz, UCB-P 071, Virlix, Zeran, Zirtec, Zirtek, Zirtin, Zyrlex, Zyrtec, Zyrzine, Cetiriz, Cetirizine dihydrochloride, Cetrak, Epirizine, Histazine-1, Tomazine |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.Cl.OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC Name | 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride |
| Show More |