missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centrin 3 Antibody (3E6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00001070-M01-50ug
This item is not returnable.
View return policy
Description
Centrin 3 Monoclonal antibody specifically detects Centrin 3 in Human, Mouse, Rat, Chicken, Zebrafish samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDown
Specifications
| Centrin 3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245 | |
| CETN3 (AAH05383, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSLALRSELVVDKTKRKKRRELSEEQKQEIKDAFELFDTDKDEAIDYHELKVAMRALGFDVKKADVLKILKDYDREATGKITFEDFNEVVTDWILERDPH | |
| 50 μg | |
| Cell Cycle and Replication | |
| 1070 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, KnockDown | |
| 3E6 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:2000, Immunoprecipitation, Knockdown Validated | |
| AAH05383 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction