Learn More
Eppendorf Centrifuge Services Preventive Maintenance
Product Code. 17913032 Shop All Eppendorf Products
Change view
Click to view available options
Service Offering:
Preventative Maintenance
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Service Offering Unit Size
17913032 Preventative Maintenance Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17913032 Supplier Eppendorf Supplier No. 0082020263
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
A schedule of planned service measures aimed at the prevention of unexpected downtimes and failures through routine maintenance and early detection of problems.
  • Cleaning of housing, condensation tray, rotor bowl and internal parts
  • Inspection of housing, lid latch, display, keypad, stability and damage
  • Testing of rotor and imbalance detection, temperature and speed sensors
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multi-purpose Centrifuges with Refrigeration
Product Type Service
Service Offering Preventative Maintenance