Learn More
Eppendorf™ Centrifuge Services - IQ/OQ - GxP
Product Code. 17349371 Shop All Eppendorf Products
Click to view available options
:
IQ/OQ GxP - Multi-purpose centrifuge w/o refrigeration
Product Code. 17349371
Brand: Eppendorf™ 0082020302
This item requires a price update prior to being ordered.
Request a Price for this item here.
GxP compliant IQ/OQ documentation support the complex GxP standards and regulations reducing your administrative workload, and provide you with qualified assurance that your instrument is installed and is operating in accordance with manufacturers' specifications.
  • Check of delivery status, and installation environment
  • Installation of instrument incl. configuration and functional check
  • Detailed IQ report
  • Test of alarm functions
  • Verification of instrument parameters
  • Detailed OQ report
  • Handling of deviation list/management
  • Signed approval by customer and service technician
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Multi-purpose centrifuge without refrigeration
Service Offering GxP Compliant IQ/OQ Documentation