Learn More
Eppendorf™ Centrifuge Adapters
Product Code. 10592774 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
1 x 15 mL Conical Tube
1 x 7 to 15 mL Blood Collection Tube
Type:
Large Bore Adapter
Small Bore Adapter
Tube Adapter
For Use With (Equipment):
Rotor A-8-17
Rotor F-35-6-30
Unit Size:
Pack of 2
Pack of 8
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Type For Use With (Equipment) unitSize
10592774 1 x 7 to 15 mL Blood Collection Tube Large Bore Adapter Rotor F-35-6-30 Pack of 2
10638004 1 x 7 to 15 mL Blood Collection Tube Small Bore Adapter Rotor F-35-6-30 Pack of 2
10789272 1 x 15 mL Conical Tube Tube Adapter Rotor A-8-17 Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10592774 Supplier Eppendorf™ Supplier No. 5427734007
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Designed for use with Eppendorf Rotors and Centrifuges
  • Accessory for Eppendorf liquid handling equipment
TRUSTED_SUSTAINABILITY
Spezifikation
Type Large Bore Adapter
Capacity 1 x 7 to 15 mL Blood Collection Tube
Max. Tube Diameter 16 mm
Max. Tube Length 90 to 105 mm
For Use With (Equipment) Rotor F-35-6-30
Description Adapter, large bore
No. per Pack 2/Pk.