Learn More
Eppendorf™ Steel Sleeves for Centrifuge Adapter
Product Code. 13246619 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
36 x 15 mL Tubes
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
13246619 36 x 15 mL Tubes Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13246619 Supplier Eppendorf™ Supplier No. 5820774002
Only null left
Add to basket
Add to basket
Designed for use with Eppendorf Rotors and Centrifuges
  • Accessory for Eppendorf liquid handling equipment
TRUSTED_SUSTAINABILITY
Specifications
Type Steel Sleeves including Adapter for Rotor
Material Steel
Capacity 36 x 15 mL Tubes
Max. Tube Diameter 16 mm
Max. Tube Length 100 mm
For Use With (Equipment) Rotor F-35-48-17
Description Steel sleeves including adapter for rotor
For Use With (Application) Blood Collection Tube
No. per Pack 24/Pk.