missing translation for 'onlineSavingsMsg'
Learn More

CENPW Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18463658 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18463658 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18463658 Supplier Novus Biologicals Supplier No. NBP281905MFV450
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CENPW Polyclonal specifically detects CENPW in Human/Mouse/Rat samples. It is validated for ELISA/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPW
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias cancer-up-regulated gene 2 protein, centromere protein W
Host Species Rabbit
Immunogen CENPW antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human CENPW. The immunogen is located within the first 50 amino acids of CENPW. Amino Acid Squence: SQRKQIKRKAPRGFLK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 387103
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.