Learn More
Greiner Bio-One CELLMASTER Polyethylene Terephthalate (PET) Roller Bottles - Short Form
Product Code. p-7075442
Click to view available options
Diameter (Metric):
116 mm
No. per Case:
1
30
Unit Size:
Pack of 18
Pack of 30
Product Code. 10218850
Brand: Greiner Bio-One 680160
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use in cell culture. Greiner Bio-One CELLMASTER Polyethylene Terephthalate (PET) Roller Bottles - Short Form is suitable for many adherent cell lines, is stable and gas permeable.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Seamless blow-moulding technique, rules out leaking seams
  • USP Class VI end product testing certified
  • Safety screw cap guarantees tightly closed, co
TRUSTED_SUSTAINABILITY
Specifications
Bottle Surface Smooth
Capacity (Metric) 2500 mL
Color Clear
Diameter (English) 4.5 in.
For Use With (Application) Cell cuture
Material Polystyrene
No. per Case 1
Sterility Sterile
Cell Growth Area 850cm2
Certified USP Class VI end product testing
Format Short form
Height (Metric) 276 mm
Product Type Roller Bottle
Height (English) 10.8 in.
Diameter (Metric) 116 mm
Closure Type Screw Cap