missing translation for 'onlineSavingsMsg'
Learn More

Cell-Surface Vimentin (CSV) Mouse anti-Human, Texas Red, Clone:84-1, Abnova™

Product Code. 16187455
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16187455 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16187455 Supplier Abnova Supplier No. H00007431MT08.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Texas Red conjugated mouse monoclonal antibody recognizes human cell-surface vimentin (CSV).

This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a dominant, pulverulent cataract

Specifications

Antigen Cell-Surface Vimentin (CSV)
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 84-1
Conjugate Texas Red
Description vimentin
Formulation PBS with no preservative; pH 7.4
Gene VIM
Gene Accession No. P08670
Gene Alias FLJ36605
Gene Symbols VIM
Host Species Mouse
Immunogen Human recombinant vimentin
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human
Content And Storage Store at 4°C in the dark. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
compliance-icons
Product Identifier
  • Cell-Surface Vimentin (CSV) monoclonal antibody, clone 84-1 (Texas Red)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P233-Keep container tightly closed.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P404-Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium phosphate monobasic, potassium phosphate dibasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.