missing translation for 'onlineSavingsMsg'
Learn More

Abcam Cell Invasion Assay Kit, Collagen I, 24-Well, 8 μm, 12 Test

Product Code. 32111579 Shop All Abcam Products
Change view
Click to view available options
Quantity:
12 Tests
Unit Size:
12 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32111579 12 Tests 12 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32111579 Supplier Abcam Supplier No. ab23588712Test
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorescent assay kit for monitoring cell invasion in suspension and adherent cells using a microplate reader.

  • Detection method: Fluorescent
  • Sample types: Suspension cells, Adherent cells
  • Suitable for use with: Microplate reader
  • Citation count: 3

The product is subject to the following: Abcam Restricted Use Statement

Specifications

For Use With (Equipment) Microplate Reader
Format 24-well Plate Kit
No. of Reactions 12 Tests
Pore Size 8 μm
Substrate Properties Protein-based Substrate
Target Molecule Collagen I
Detection Method Fluorescence
Sample Type Suspension cells, Adherent cells
Quantity 12 Tests
For Use With (Application) Cell Invasion Assay, Functional Studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.