Learn More
Cefotetan disodium, Thermo Scientific™
A broad spectrum antibiotic
119.00€
Chemical Identifiers
| CAS | 74356-00-6 |
|---|---|
| Molecular Formula | C17H15N7Na2O8S4 |
| Molecular Weight (g/mol) | 619.568 |
| MDL Number | MFCD00941449 |
| InChI Key | ZQQALMSFFARWPK-ZTQQJVKJSA-L |
| PubChem CID | 53024 |
| IUPAC Name | disodium;(6R,7S)-7-[[4-(2-amino-1-carboxylato-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| SMILES | CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)[O-])S4)OC)SC2)C(=O)[O-].[Na+].[Na+] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 74356-00-6 | |
| 619.568 | |
| ZQQALMSFFARWPK-ZTQQJVKJSA-L | |
| disodium;(6R,7S)-7-[[4-(2-amino-1-carboxylato-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| C17H15N7Na2O8S4 | |
| MFCD00941449 | |
| 53024 | |
| CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)[O-])S4)OC)SC2)C(=O)[O-].[Na+].[Na+] |
Specifications
| 74356-00-6 | |
| C17H15N7Na2O8S4 | |
| 798°C | |
| ZQQALMSFFARWPK-ZTQQJVKJSA-L | |
| disodium;(6R,7S)-7-[[4-(2-amino-1-carboxylato-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
| 53024 | |
| Cefotetan disodium |
| White | |
| MFCD00941449 | |
| Freely soluble in aqueous solution | |
| CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)[O-])S4)OC)SC2)C(=O)[O-].[Na+].[Na+] | |
| 619.568 | |
| 619.58 |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334
missing translation for 'einecsNumber' : 277-834-9
missing translation for 'rtecsNumber' : XI0330600
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.