CAS RN 74356-00-6
CAS RN 74356-00-6
IUPAC Name:
disodium (6R,7S)-7-{4-[carbamoyl(carboxylato)methylidene]-1,3-dithietane-2-amido}-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms:
cefotetan disodium cefotetan sodium
Molecular Weight (g/mol):
619.57
Molecular Formula:
C17H15N7Na2O8S4
InChi Key:
ZQQALMSFFARWPK-GLHLDKNHSA-L
SMILES:
[Na+].[Na+].CO[C@]1(NC(=O)C2SC(S2)=C(C(N)=O)C([O-])=O)[C@H]2SCC(CSC3=NN=NN3C)=C(N2C1=O)C([O-])=O
1
–
1
of
1
results
1
Cefotetan disodium, Thermo Scientific™
CAS: 74356-00-6 Molecular Formula: C17H15N7Na2O8S4 Molecular Weight (g/mol): 619.568 MDL Number: MFCD00941449 InChI Key: ZQQALMSFFARWPK-ZTQQJVKJSA-L PubChem CID: 53024 IUPAC Name: disodium;(6R,7S)-7-[[4-(2-amino-1-carboxylato-2-oxoethylidene)-1,3-dithietane-2-carbonyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate SMILES: CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C4SC(=C(C(=O)N)C(=O)[O-])S4)OC)SC2)C(=O)[O-].[Na+].[Na+]