Learn More
Cefdinir, 95%, Thermo Scientific™
157.00€ - 474.00€
Chemical Identifiers
| CAS | 91832-40-5 |
|---|---|
| Molecular Formula | C14H13N5O5S2 |
| Molecular Weight (g/mol) | 395.408 |
| MDL Number | MFCD00865030 |
| InChI Key | RTXOFQZKPXMALH-GHXIOONMSA-N |
| PubChem CID | 6915944 |
| ChEBI | CHEBI:3485 |
| IUPAC Name | (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
| SMILES | C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15447809
|
Thermo Scientific Alfa Aesar
J66447.03 |
1g |
157.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15457809
|
Thermo Scientific Alfa Aesar
J66447.06 |
5g |
474.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- A broad spectrum antibiotic targeting both gram positive and gram negative pathogens
Chemical Identifiers
| 91832-40-5 | |
| 395.408 | |
| RTXOFQZKPXMALH-GHXIOONMSA-N | |
| CHEBI:3485 | |
| C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O |
| C14H13N5O5S2 | |
| MFCD00865030 | |
| 6915944 | |
| (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
Specifications
| 91832-40-5 | |
| C14H13N5O5S2 | |
| Soluble in water | |
| C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O | |
| 395.408 | |
| CHEBI:3485 | |
| 95% |
| White to Yellow | |
| MFCD00865030 | |
| RTXOFQZKPXMALH-GHXIOONMSA-N | |
| (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid | |
| 6915944 | |
| 395.41 | |
| Cefdinir |
Safety and Handling
missing translation for 'rtecsNumber' : XI0367250
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold