missing translation for 'onlineSavingsMsg'
Learn More

Cefdinir, 95%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J66447.06

Quantity 5g

 View more versions of this product

Product Code. 15457809

  • 474.00€ / 5g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • A broad spectrum antibiotic targeting both gram positive and gram negative pathogens
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

91832-40-5
395.408
RTXOFQZKPXMALH-GHXIOONMSA-N
CHEBI:3485
C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O
C14H13N5O5S2
MFCD00865030
6915944
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

Specifications

91832-40-5
5g
MFCD00865030
RTXOFQZKPXMALH-GHXIOONMSA-N
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
6915944
395.41
Cefdinir
White to Yellow
C14H13N5O5S2
Soluble in water
C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O
395.408
CHEBI:3485
95%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.