accessibility menu, dialog, popup

UserName

CAS RN 91832-40-5

H2NSNNOHOHN(R)(R)SNOHOOCH2(Z)

CAS RN 91832-40-5

IUPAC Name: (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(N-hydroxyimino)acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: cefdinir cefdinir product abbott brand of cefdinir cefdinirum cefdinir product (product) cefdinyl cefdinir preparation cefdirnir CFDN cefdinir (substance)
Molecular Weight (g/mol): 395.41
Molecular Formula: C14H13N5O5S2
InChi Key: RTXOFQZKPXMALH-GHXIOONMSA-N
SMILES: NC1=NC(=CS1)C(=N\O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)