missing translation for 'onlineSavingsMsg'
Learn More

CEBP alpha Antibody (15C8), Novus Biologicals™

Product Code. 18537030 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18537030 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18537030 Supplier Novus Biologicals Supplier No. NB60014380.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

CEBP alpha Monoclonal specifically detects CEBP alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEBP alpha
Applications Western Blot
Classification Monoclonal
Clone 15C8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Flow Cytometry 3 - 5 μg/ 106 cells, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Gene Accession No. P49715
Gene Alias C/EBP alpha, C/EBP-alpha, CCAAT/enhancer binding protein (C/EBP), alpha, CCAAT/enhancer-binding protein alpha, CEBP
Gene Symbols CEBPA
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues M(1) E S A D F Y E V E P R P P(14) of mouse C/EBP alpha.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1050
Test Specificity Expected to cross-react with Xenopus laevis (100% identity with immunogen), Zebrafish (92% identity with immunogen), Aspergillus nidulans (85% identity with immunogen), Goldfish (92% identity with immunogen), Hamster (100% identity with immunogen), Neurospora crassa (85% identity with immunogen) and S. pombe (92% identity with immunogen) due to sequence homology. Not yet tested in other species.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.