missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CEACAM8 (Human) Recombinant Protein
Human CEACAM8 full-length ORF ( AAH26263, 1 a.a. - 349 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001088-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGPISAPSCRWRIPWQGLLLTASLFTFWNPPTTAQLTIEAVPSNAAEGKEVLLLVHNLPQDPRGYNWYKGETVDANRRIIGYVISNQQITPGPAYSNRETIYPNASLLMRNVTRNDTGSYTLQVIKLNLMSEEVTGQFSVHPETPKPSISSNNSNPVEDKDAVAFTCEPETQNTTYLWWVNGQSLPVSPRLQLSNGNRTLTLLSVTRNDVGPYECEIQNPASANFSDPVTLNVLYGPDAPTISPSDTYYHAGVNLNLSCHAASNPPSQYSWSVNGTFQQYTQKLFIPNITTKNSGSYACHTTNSATGRNRTTVRMITVSDALVQGSSPGLSARATVSIMIGVLARVALI
Specifications
AAH26263 | |
carcinoembryonic antigen-related cell adhesion molecule 8 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CEACAM8 | |
GST |
1088 | |
Wheat germ expression system | |
25 μg | |
CD66b, CD67, CGM6, NCA-95 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |