Learn More
CEACAM6/CD66c Antibody (01), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89670H
Product Code. 18642590
  • 660.00€ / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
CEACAM6/CD66c Monoclonal antibody specifically detects CEACAM6/CD66c in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEACAM6/CD66c
Monoclonal
HRP
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA
01
ELISA
carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM6/CD66c (NP_002474.3; Met1-Gly320).
0.1 mL
Cancer
4680
Store at 4°C in the dark.