missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Rabbit anti-Human, Clone: CDX2/4394R, Novus Biologicals™
Rabbit Monoclonal Antibody
551.00€
Specifications
| Antigen | CDX2 |
|---|---|
| Clone | CDX2/4394R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
CDX2 Monoclonal specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| CDX2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 1045 | |
| IgG | |
| Protein A or G purified |
| CDX2/4394R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
| Recombinant fragments and synthetic peptides from human CDX2 protein (exact sequences are proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title