missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDO Monoclonal Antibody (274133)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561153
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
Specifications
| CDO | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q32MD9 | |
| CDON | |
| Mouse myeloma cell line NS0-derived recombinant mouse CDO, Val22-Leu963. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 274133 | |
| Unconjugated | |
| CDON | |
| CAM-related/down-regulated by oncogenes; Cd0; Cdo; CDON; Cdon homolog; CDON1; cell adhesion associated, oncogene regulated; cell adhesion molecule-related/down-regulated by oncogenes; HPE11; immunoglobulin superfamily member; immunoglobulin superfamily member CDO; oncogene-regulated cell adhesion molecule; ORCAM; surface glycoprotein, Ig superfamily member | |
| Rat | |
| Protein A/G | |
| RUO | |
| 57810 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction