missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP2 Antibody (CL3392) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59027AF532
This item is not returnable.
View return policy
Description
CDK5RAP2 Monoclonal antibody specifically detects CDK5RAP2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| CDK5RAP2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| C48, CDK5 activator-binding protein C48, CDK5 regulatory subunit associated protein 2, CDK5 regulatory subunit-associated protein 2, centrosomal protein 215 kDa, Centrosome-associated protein 215, Cep215, DKFZp686B1070, DKFZp686D1070, FLJ10867, KIAA1633microcephaly, primary autosomal recessive 3, MCPH3 | |
| This antibody was developed using a recombinant protein derived from Q96SN8, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Markers, Chromatin Research | |
| 55755 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL3392 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction