missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 624.00€
Specifications
| Antigen | CDK5RAP1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18288364
|
Novus Biologicals
NBP2-55839 |
100 μL |
624.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605308
|
Novus Biologicals
NBP2-55839-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDK5RAP1 Polyclonal specifically detects CDK5RAP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| CDK5RAP1 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 51654 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RLKDDVPEEVKLRRLEELITIFREEATKANQTSVGCTQLVLVEGLSKRSATDLCGRNDGNLKVIFPDAEMEDVNNPGLRVRAQPGDYVLVKITSAS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| C20orf34, C42, CDK5 activator-binding protein C42, CDK5 regulatory subunit associated protein 1, CDK5 regulatory subunit-associated protein 1, CDK5RAP1.3, CDK5RAP1.4, CGI-05, chromosome 20 open reading frame 34, HSPC167 | |
| CDK5RAP1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title