missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK4 Antibody (8H4), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | CDK4 |
|---|---|
| Clone | 8H4 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043048
|
Novus Biologicals
NBP3-26665-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043956
|
Novus Biologicals
NBP3-26665-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDK4 Monoclonal antibody specifically detects CDK4 in Human samples. It is validated for Western Blot, ELISASpecifications
| CDK4 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4 | |
| A synthesized peptide derived from Human CDK4 [UniProt P11802] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8H4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Protein Kinase, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1019 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title