missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCP1 Antibody (30D4), Novus Biologicals™
Human Monoclonal Antibody
343.00€ - 533.00€
Spezifikation
| Antigen | CDCP1 |
|---|---|
| Clone | 30D4 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30043562
|
Novus Biologicals
NBP3-26667-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043555
|
Novus Biologicals
NBP3-26667-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
CDCP1 Monoclonal antibody specifically detects CDCP1 in Human, Mouse, Cynomolgus Monkey samples. It is validated for ELISASpezifikation
| CDCP1 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Cynomolgus Monkey | |
| CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
| Recombinant Human CDCP1 protein [UniProt Q9H5V8] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 30D4 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Phospho Specific | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 64866 | |
| IgG1 | |
| Affinity purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts