missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74621JF646
This item is not returnable.
View return policy
Description
CDCA3 Polyclonal antibody specifically detects CDCA3 in Human samples. It is validated for Western Blot
Specifications
| CDCA3 | |
| Polyclonal | |
| Western Blot | |
| C8, cell division cycle associated 3, FBXO52, Gene-rich cluster protein C8, GRCC8cell division cycle-associated protein 3, MGC2577, TOME1, TOME-1gene rich cluster, C8, trigger of mitotic entry 1, Trigger of mitotic entry protein 1 | |
| The immunogen recognized by this antibody maps to a region between residue 200 and 230 of human cell division cycle associated 3 (trigger of mitotic entry 1) using the numbering given in entry NP_112589.1 (GeneID 83461). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 83461 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction