missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc27 Antibody (AF 3.1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2606IR
This item is not returnable.
View return policy
Description
Cdc27 Monoclonal specifically detects Cdc27 in Mammal, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications
| Cdc27 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CDC27 | |
| A peptide corresponding to the C-terminal 10 residues of human cdc27 coupled to KLH. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 996 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| AF 3.1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| ANAPC3CDC27 homolog, anaphase promoting complex subunit 3, Anaphase-promoting complex subunit 3, anaphase-promoting complex, protein 3, APC 3, APC3cell division cycle 27, CDC27Hs, cell division cycle 27 homolog (S. cerevisiae), cell division cycle protein 27 homolog, D0S1430E, D17S978E, HNUC, H-NUC, NUC2, nuc2 homolog | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mammalia, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction