missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc20 Antibody (BA8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
201.00€
Specifications
| Antigen | Cdc20 |
|---|---|
| Clone | BA8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, CyTOF-ready |
| Applications | Western Blot |
Description
Cdc20 Monoclonal specifically detects Cdc20 in Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Cdc20 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| CDC20 cell division cycle 20 homolog, CDC20 cell division cycle 20 homolog (S. cerevisiae), cell division cycle 20 homolog (S. cerevisiae), cell division cycle protein 20 homolog, MGC102824, p55CDCS. cerevisiae, homolog) | |
| CDC20 | |
| IgG2a | |
| Protein A purified | |
| Cdc20 (BA8). Cross-reacts with Xenopus laevis. Not yet tested in other species. |
| BA8 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Xenopus species | |
| 991 | |
| Peptide from the carboxy terminus of the protein. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title