missing translation for 'onlineSavingsMsg'
Learn More

CDC2/CDK1, p Thr161 Antibody, Novus Biologicals™

Product Code. 18228921 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
Unit Size:
0.05mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18228921 0.025 mL 25µL
18207542 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18228921 Supplier Novus Biologicals Supplier No. NB100818370.025ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC2/CDK1 Polyclonal specifically detects CDC2/CDK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC2/CDK1 (p Thr161)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Formulation phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Gene Accession No. P06493
Gene Alias CDC28A, CDC2MGC111195, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2
Gene Symbols CDK1
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human CDC2 around the phosphorylation site of threonine161 (T-Y-TP-H-E).
Molecular Weight of Antigen 34 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 983
Test Specificity CDC2 (specific to Phospho-Thr161) detects endogenous levels of CDC2 only when phosphorylated at threonine 161.
Target Species Rat, Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.