Learn More
CD90/Thy1 Antibody (1aG4/C5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-44877CL1
This item is not returnable.
View return policy
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human (Negative),Mouse,Rat (Negative) samples. It is validated for Flow Cytometry,Western Blot,Functional Assay,Flow (Cell Surface),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| CD90/Thy1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| thymocytes from C3H/Di mice (alelic form: Thy-1.2 positive) | |
| 0.1 mL | |
| Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| 7070 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG3 |
| Flow Cytometry, Western Blot, Functional Assay, Flow Cytometry (Cell Surface), CyTOF | |
| 1aG4/C5 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human (Negative), Mouse, Rat (Negative) | |
| Purified |