Learn More
CD9 Mouse anti-Human, Brilliant Violet 786, Clone: M-L13, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 743051
Description
The M-L13 monoclonal antibody specifically binds to a 24 kDa type III transmembrane protein that is expressed on platelets, pre-B cells, monocytes, endothelial and epithelial cells. CD9 belongs to a family of membrane proteins called tetraspanins that transverse the membrane four times. CD9 is weakly expressed on resting mature B cells. M-L13 induces platelet aggregation and activation. This antibody is also suitable for staining acetone-fixed, frozen tissue sections.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
Specifications
| CD9 | |
| M-L13 | |
| Brilliant Violet 786 | |
| CD9 antigen (p24), 5H9, BA2, BTCC-1, DRAP-27, GIG2, MIC3, MRP-1, TSPAN29 | |
| Human C-ALL Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.