missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Antibody (HI9a), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18795AF532
This item is not returnable.
View return policy
Description
CD9 Monoclonal antibody specifically detects CD9 in Human, Bovine, Canine, Equine, Monkey, Rabbit, Sheep samples. It is validated for Flow Cytometry
Specifications
| CD9 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
| A recombinant protein of human CD9 | |
| 0.1 mL | |
| Cancer, Stem Cell Markers | |
| 928 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| HI9a | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Bovine, Canine, Equine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction