missing translation for 'onlineSavingsMsg'
Learn More

CD8 Mouse anti-Human, PE, Clone: HIT8A, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555635

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 15878318

  • 435.00€ / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

Reacts with the α subunit (32kDa) of the two-chain complex. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. CD8 is expressed on T cytotoxic/suppressor cell populations. HIT8a stains approximately 13 - 48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. Clones HIT8a and RPA-T8 are not cross-blocking.

Host Species: Mouse
Clone: HIT8a
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

Specifications

Specifications

CD8
Monoclonal
PE
Mouse
100 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
HIT8A
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.