missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human, PE, Clone: HIT8A, BD
Mouse Monoclonal Antibody
185.00€ - 435.00€
Specifications
Antigen | CD8 |
---|---|
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15820639
|
BD Biosciences
560959 |
25 Tests |
185.00€
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15878318
|
BD Biosciences
555635 |
100 Tests |
435.00€
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Reacts with the α subunit (32 kDa) of the two-chain complex. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. CD8 is expressed on T cytotoxic/suppressor cell populations. HIT8a stains approximately 13 - 48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. Clones HIT8a and RPA-T8 (Cat. No. 555364) are not cross-blocking.
Specifications
CD8 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.