missing translation for 'onlineSavingsMsg'
Learn More

CD8 Mouse anti-Human, PE, Clone: HIT8A, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560959

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15820639

  • 185.00€ / Each
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

Reacts with the α subunit (32 kDa) of the two-chain complex. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. CD8 is expressed on T cytotoxic/suppressor cell populations. HIT8a stains approximately 13 - 48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. Clones HIT8a and RPA-T8 (Cat. No. 555364) are not cross-blocking.

Specifications

Specifications

CD8
Monoclonal
PE
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
HIT8a
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.