missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (YTS 105.18) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
291.00€ - 558.00€
Specifications
| Antigen | CD8 alpha |
|---|---|
| Clone | YTS 105.18 |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry-Frozen 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Blocking Assay, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18788943
|
Novus Biologicals
NBP2-52659-0.025MG |
0.025 mg |
291.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18200571
|
Novus Biologicals
NBP2-52659-0.2MG |
0.2 mg |
558.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8 Monoclonal specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence, CyTOF-ready, Block/Neutralize.Specifications
| CD8 alpha | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry-Frozen 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD8, CD8 antigen, alpha polypeptide (p32), CD8a antigen, CD8a molecule, Leu2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, RPA-T8, RPA-T8 antibody flow, RPA-T8 CD8, RPA-T8 Clone, RPA-T8 Flow, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2 | |
| CD8A | |
| IgG | |
| Protein A or G purified | |
| Recognizes a non polymorphic epitope on the mouse CD8 alpha chain. This antibody has been reported to block MHC I dependent T cell responses in vitro and in vivo. |
| YTS 105.18 | |
| Flow Cytometry, Immunohistochemistry (Frozen), Blocking Assay, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| 925 | |
| Ly-2-transfected mouse L cells. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title