missing translation for 'onlineSavingsMsg'
Learn More

CD79B Mouse anti-Human, Clone: rIGB/1842, Novus Biologicals™

Mouse Monoclonal Antibody

499.00€

Specifications

Antigen CD79B
Clone rIGB/1842
Concentration 0.2 mg/mL
Dilution ELISA 2 to 4 μg/mL
Applications ELISA
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18627316
View Documents
Novus Biologicals
NBP3-07439-100ug
100 μg
499.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD79B Monoclonal specifically detects CD79B in Human samples. It is validated for ELISA.
Specifications

Specifications

CD79B
0.2 mg/mL
ELISA
Unconjugated
Mouse
Human
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Recombinant human CD79B protein fragment (around aa29-159) (exact sequence is proprietary)
Primary
Store at 4°C.
rIGB/1842
ELISA 2 to 4 μg/mL
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
974
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.