missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD74 Monoclonal antibody specifically detects CD74 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CD74 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rCLIP/8679 |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33 |
| Host Species | Mouse |
| Immunogen | Recombinant fragment of human CD74 protein (exact sequence is proprietary) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?