missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68, Mouse anti-Human, Clone: PM-1K, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB1715.50ug
This item is not returnable.
View return policy
Specifications
| CD68 | |
| Monoclonal | |
| Unconjugated | |
| In PBS (0.1% proclin, 2.0% Block Ace) | |
| DKFZp686M18236/GP110/SCARD1 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Guinea Pig, Human | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Frozen) | |
| PM-1K | |
| Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| CD68 molecule | |
| CD68 | |
| Native purified from human peritoneal cells from patients with endometriosis. | |
| RUO | |
| 968 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction