missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD68 Monoclonal Antibody (eBioY1/82A (Y1/82A)), Alexa Fluor™ 647, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
351.00€
Specifications
| Antigen | CD68 |
|---|---|
| Clone | eBioY1/82A (Y1/82A) |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16625444
![]() |
Invitrogen™
51-0689-42 |
100 Tests | |||||||
Description
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.Specifications
| CD68 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen | |
| Cd68 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Cd68 |
| eBioY1/82A (Y1/82A) | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Mouse | |
| Human | |
| P34810 | |
| 968 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title