missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CD63 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | LAMP3/9191R |
| Conjugate | mFluor Violet 500 SE |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 |
| Host Species | Rabbit |
| Immunogen | Recombinant fragment (around aa100-200) of human CD63 protein (exact sequence is proprietary) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?