missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LILRC2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148820
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
LILRC2, also known as Leukocyte Immunoglobulin-Like Receptor Subfamily C Member 2 (LILRC2), is a gene that encodes a member of the leukocyte immunoglobulin-like receptor (LILR) family. These receptors are primarily expressed on the surface of various immune cells, including B cells, dendritic cells, monocytes, and natural killer (NK) cells. LILRC2 functions as an inhibitory receptor, playing a critical role in the modulation of immune responses. By binding to specific ligands, such as major histocompatibility complex (MHC) class I molecules, LILRC2 can transmit inhibitory signals that dampen immune cell activation, thereby maintaining immune homeostasis and preventing excessive or inappropriate immune responses. This regulatory function is crucial for preventing autoimmunity and maintaining tolerance to self-antigens. Dysregulation or altered expression of LILRC2 has been associated with various immune-related conditions, including autoimmune diseases, chronic infections, and cancers. Understanding the specific roles and signaling mechanisms of LILRC2 is important for developing therapeutic strategies aimed at modulating immune responses, enhancing immune tolerance, and treating immune-mediated diseases. Research into LILRC2 provides valuable insights into the intricate balance of immune regulation and the potential for targeting inhibitory receptors in immunotherapy.
Specifications
| LILRC2 | |
| Polyclonal | |
| Unconjugated | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Mouse myeloma cell line NS0-derived recombinant rat LILRC2, Gly24-Asn230. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction