missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a member of the immunoglobulin superfamily. The encoded protein is a ligand of the T lymphocyte CD2 protein, and functions in adhesion and activation of T lymphocytes. The protein is localized to the plasma membrane. Alternatively spliced transcript variants have been described.
Specifications
Specifications
| Antigen | CD58 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | CD58 |
| Gene Accession No. | P19256 |
| Gene Alias | Ag3, CD58, CD58 molecule, LFA 3, LFA3, Surface glycoprotein LFA 3 |
| Gene Symbols | CD58 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?