missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Antibody (NK1/7565), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20869UV
619.65 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD57 | |
| Monoclonal | |
| DyLight 350 | |
| HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7 | |
| Recombinant fragment (around aa1-200) of human CD57 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cell Biology, Immunology | |
| 27087 | |
| Store at 4°C in the dark. | |
| IgG2 λ |
| Immunohistochemistry (Paraffin) | |
| NK1/7565 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction