missing translation for 'onlineSavingsMsg'
Learn More

CD56 / NCAM1 / NKH1 Mouse, Clone: NCAM1/784, enQuireBio™

Product Code. 15970558
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15970558 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15970558 Supplier EnquireBio Supplier No. 4684MSM4P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen CD56 / NCAM1 / NKH1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Cellular Localization Cell surface
Classification Monoclonal
Clone NCAM1/784
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.05% BSA and 0.05% azide
Gene Accession No. NP_000606
Gene Symbols NCAM1
Host Species Mouse
Immunogen Recombinant human NCAM1 protein
Molecular Weight of Antigen 180, 145, 125kDa
Positive Control Cerebellum, Pancreas, Neuroblastoma
Purification Method Protein A/G Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4684
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.