missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Antibody (OX-101) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66632-0.025MG
This item is not returnable.
View return policy
Description
CD47 Monoclonal specifically detects CD47 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.
Specifications
| CD47 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 | |
| P97829 | |
| CD47 | |
| Rat thymocytes | |
| 0.025 mg | |
| Immunology, Signal Transduction | |
| 961 | |
| Rat | |
| Supernatant |
| Western Blot, Immunoprecipitation, Flow Cytometry | |
| OX-101 | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Antibody recognizes the rat homologue of human CD47, which has been identified as the ligand for the rat signal regulatory protein (SIRP). Rat CD47 is expressed primarily by lymphocytes and macrophages but has also been detected at high levels on brain endothelial cells. Clone OX-101 has been reported to block the interaction of CD47 and SIRP. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction