Specifications
| Product Type | Non-magnetic Cell Isolation Kit |
| Content And Storage | 2°C to 8°C |
| Donor Source | Human Whole Blood or Buffy Coat |
| For Use With (Application) | Isolation of label-free cells |
| Sufficient For | 9 mL Whole Blood |
| Quantity | 1 Kit, Human |