Learn More
CD45RA/CD45RO/CD3/CD8 Mouse anti-Human, APC,FITC,PE,PerCP, Clone: SK1,L48,UCHL1,SK7 (also known as Leu-4), BD Multitest™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 340574
Product Code. 15855928
  • 1040.00€ / Each
Description
CD45RA/CD45RO/CD3/CD8 Monoclonal antibody specifically detects CD45RA/CD45RO/CD3/CD8 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD45RA/CD45RO/CD3/CD8
Monoclonal
APC/FITC/PE/PerCP
50 Tests
Primary
Refrigerate (2 to 8°C)
IgG1 κ, IgG2a κ
Flow Cytometry
SK1, L48, UCHL1, SK7 (also known as Leu-4)