Description
The CD45 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human CD45. This antibody recognizes human, rat antigen. The CD45 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
Specifications
| Antigen | CD45 |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry |
| Clone | 6O19 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 5788 |
| Target Species | Human, Rat |
| Isotype | IgG |