missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 FITC/CD34 PE Mouse anti-Human, FITC,PE, Clone: 2D1,8G12 (also known as HPCA2), BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 341071
This item is not returnable.
View return policy
Description
CD45 FITC/CD34 PE Monoclonal antibody specifically detects CD45 FITC/CD34 PE in Human samples. It is validated for Flow Cytometry.
Specifications
| CD45 FITC/CD34 PE | |
| Monoclonal | |
| FITC/PE | |
| 50 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
| Flow Cytometry | |
| 2D1, 8G12 (also known as HPCA2) | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction